Stopcocks, three way
1,5 mm, 14,5 NS
Katalogové číslo 227-3231
Dodavatel: Lenz Laborglas
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys with T-bore
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
DIN 12554
Documents
BSE TSE Glass Columns
Certifikáty BSE/TSE
Specifikace
- Ø otvoru:1,5 mm
- Kohout:14,5 NS