Orders LinkContactUsLinkComponent
Stopcocks, two-way
Stopcocks, two-way
4,0 mm, 18,8 NS
Catalog # LENZ02431804
Supplier:   Lenz Laborglas
Stopcocks, two-way
Catalog # LENZ02431804
Supplier:  Lenz Laborglas
Stopcocks, two-way
Stopcocks, two-way
4,0 mm, 18,8 NS
Price
2 960,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stopcocks are made of DURAN® borosilicate glass with solid NS glass plug, screwthread retaining device, and two oblique bores.
  • Excellent chemical resistance
  • Excellent temperature resistance
With two oblique bores.
Leakage rate accoring to DIN 12540-1.
Documents
Specifications
  • Ø otvoru:
    4,0 mm
  • Kohout:
    18,8 NS