Orders LinkContactUsLinkComponent
Stopcocks, two-way
Stopcocks, two-way
2,5 mm, 14,5 NS
Katalogové číslo LENZ02431402
Dodavatel:   Lenz Laborglas
Stopcocks, two-way
Katalogové číslo LENZ02431402
Dodavatel:  Lenz Laborglas
Stopcocks, two-way
Stopcocks, two-way
2,5 mm, 14,5 NS
Přejít na:
Price
2 320,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stopcocks are made of DURAN® borosilicate glass with solid NS glass plug, screwthread retaining device, and two oblique bores.
  • Excellent chemical resistance
  • Excellent temperature resistance
With two oblique bores.
Leakage rate accoring to DIN 12540-1.
Documents
Specifikace
  • Ø otvoru:
    2,5 mm
  • Kohout:
    14,5 NS