Stopcocks, two-way
4,0 mm, 18,8 NS
LENZ02401804
Stopcocks are made of DURAN® borosilicate glass with solid NS glass plug, screwthread retaining device, and two oblique bores.
- Excellent chemical resistance
- Excellent temperature resistance
With two oblique bores.
Leakage rate accoring to DIN 12540-1.
- Ø otvoru:4,0 mm
- Kohout:18,8 NS