Orders LinkContactUsLinkComponent
Stopcocks, one-way
17, 4,0 mm, 18,8 NS
Katalogové číslo 201-3116
Dodavatel:   Lenz Laborglas
Stopcocks, one-way
Katalogové číslo 201-3116
Dodavatel:  Lenz Laborglas
Stopcocks, one-way
Stopcocks, one-way
17, 4,0 mm, 18,8 NS
Přejít na:
Price
1 360,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
 : DIN 12554.
Specifikace
  • Ø otvoru:
    4,0 mm
  • Sekvence:
    17
  • Kohout:
    18,8 NS