Orders LinkContactUsLinkComponent
Stopcocks, one-way
13, 2,5 mm, 12,5 NS
Katalogové číslo 201-0494
Dodavatel:   Lenz Laborglas
Stopcocks, one-way
Katalogové číslo 201-0494
Dodavatel:  Lenz Laborglas
Stopcocks, one-way
Stopcocks, one-way
13, 2,5 mm, 12,5 NS
Přejít na:
Price
885,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifications: DIN 12554.
Specifikace
  • Ø otvoru:
    2,5 mm
  • Sekvence:
    13
  • Kohout:
    12,5 NS