Orders LinkContactUsLinkComponent
Stopcocks, one-way
Stopcocks, one-way
11, 1,5 mm, 12,5 NS
  201-1106
Stopcocks, one-way
  201-1106
Stopcocks, one-way
Stopcocks, one-way
11, 1,5 mm, 12,5 NS
842,00 Kč
1 * 1 KS
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifications: DIN 12554.
Documents
  • Ø otvoru:
    1,5 mm
  • Sekvence:
    11
  • Kohout:
    12,5 NS