Orders LinkContactUsLinkComponent
Stopcocks, one-way
06, 4,0 mm, 14,5 NS
  227-3135
Stopcocks, one-way
  227-3135
Stopcocks, one-way
Stopcocks, one-way
06, 4,0 mm, 14,5 NS
851,00 Kč
1 * 1 KS
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
 : DIN 12554.
  • Ø otvoru:
    4,0 mm
  • Sekvence:
    06
  • Kohout:
    14,5 NS