Orders LinkContactUsLinkComponent
Spherical ground joints
Spherical ground joints
, , , 100 mm, -, S 13/5
Katalogové číslo 201-4212
Dodavatel:   Lenz Laborglas
Spherical ground joints
Katalogové číslo 201-4212
Dodavatel:  Lenz Laborglas
Spherical ground joints
Spherical ground joints
, , , 100 mm, -, S 13/5
Přejít na:
Price
301,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
Spherical cup and ball joint with inch markings. Fused edges. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving relatively high resistance to temperature changes.
DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641
Certifications: DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641.
Documents
Specifikace
  • Kulička:
    -
  • Zásuvka:
    S 13/5
  • Celková délka:
    100 mm