Orders LinkContactUsLinkComponent
Lifting platforms for Lauda baths
03, Lifting platform, for 1× E 10/E 15/ET 12/E 112 T/E 212 T/E 312/E 111/E 211/ET 12/ET 20 and 2× A 24/E 20/E 25/E 120 T/E 220 T/ET 20/E 119/E 219/E 125/E 225/E 320/E 326, 140×270 mm
Katalogové číslo 461-2909
Dodavatel:   LAUDA
Lifting platforms for Lauda baths
Katalogové číslo 461-2909
Dodavatel:  LAUDA
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
03, Lifting platform, for 1× E 10/E 15/ET 12/E 112 T/E 212 T/E 312/E 111/E 211/ET 12/ET 20 and 2× A 24/E 20/E 25/E 120 T/E 220 T/ET 20/E 119/E 219/E 125/E 225/E 320/E 326, 140×270 mm
Přejít na:
Price
11 080,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These platforms are made from stainless steel. For use with Lauda baths.
  • Height adjustable in eight steps
  • To be fixed on one side of the bath
  • Not for use with bath cover or gable cover
Specifikace
  • Popis:
    Lifting platform, for 1× E 10/E 15/ET 12/E 112 T/E 212 T/E 312/E 111/E 211/ET 12/ET 20 and 2× A 24/E 20/E 25/E 120 T/E 220 T/ET 20/E 119/E 219/E 125/E 225/E 320/E 326
  • D׊:
    140×270 mm
  • Sekvence:
    03