Orders LinkContactUsLinkComponent
Decontamination solution, DNA AWAY®
Decontamination solution, DNA AWAY®
01, DNA AWAY®, 250 mL
Katalogové číslo 732-2353
   Low Manufacturing Impact
Decontamination solution, DNA AWAY®
Katalogové číslo 732-2353
   Low Manufacturing Impact
Číslo CAS:  
Decontamination solution, DNA AWAY®
Decontamination solution, DNA AWAY®
01, DNA AWAY®, 250 mL
Přejít na:
Číslo CAS:  
Price
549,00 Kč
Akce
Platné do 31/03/2026
Přihlásit se pro dostupnost
1 * 250 mL
Product Details & Documents
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers and more.
  • Suitable for PCR applications, preventing false-positive PCR results
  • Easy cleaning of various types of surfaces
Specifikace
Specifikace
  • Bal.:
    250 mL
  • Popis:
    DNA AWAY®
  • Sekvence:
    01
  • Cat. No.:
    732-2353
  • Certifikát ochrany životního prostředí:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact