Dewar flasks, type GSS
08, Loose lid insulating plug, GSS 1000
Katalogové číslo 478-5143
Dodavatel: KGW
Přejít na:
Product Details & Documents
Dewar flasks in stainless steel for storage and transport of liquid nitrogen, dry ice and other coolants. They are vacuum insulated and have a temperature range from −200 to +200 °C.
- With side grip and transport handle (except model GSS 500)
Specifikace
- Popis:Loose lid insulating plug
- Sekvence:08
- Určeno pro:GSS 1000