Orders LinkContactUsLinkComponent
Spherical Dewar flasks, type AL
Spherical Dewar flasks, type AL
07, Glass refill, 24 A, Spherical dewar flask, 24 AL
Catalog # 478-4408
Supplier:   KGW
Spherical Dewar flasks, type AL
Catalog # 478-4408
Supplier:  KGW
Spherical Dewar flasks, type AL
Spherical Dewar flasks, type AL
07, Glass refill, 24 A, Spherical dewar flask, 24 AL
Price
18 740,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Dewar flasks made from silvered borosilicate glass, brushed aluminium protective covering.
  • With lid and handle
Specifications
  • Popis:
    Glass refill, 24 A
  • Sekvence:
    07
  • Určeno pro:
    Spherical dewar flask, 24 AL