Orders LinkContactUsLinkComponent
Spherical Dewar flasks, type AL
Spherical Dewar flasks, type AL
05, Glass refill, 22 A, Spherical dewar flask, 22 AL
Catalog # 478-0735
Supplier:   KGW
Spherical Dewar flasks, type AL
Catalog # 478-0735
Supplier:  KGW
Spherical Dewar flasks, type AL
Spherical Dewar flasks, type AL
05, Glass refill, 22 A, Spherical dewar flask, 22 AL
Price
9 820,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Dewar flasks made from silvered borosilicate glass, brushed aluminium protective covering.
  • With lid and handle
Specifications
  • Popis:
    Glass refill, 22 A
  • Sekvence:
    05
  • Určeno pro:
    Spherical dewar flask, 22 AL