Orders LinkContactUsLinkComponent
Pipette tips
42, Standard, 250 - 1000 µl, -, Transparent, Beckman®, Sherwood®, Lancer®, Bulk
  613-0500
 :   KARTELL
Pipette tips
  613-0500
 :  KARTELL
Pipette tips
Pipette tips
42, Standard, 250 - 1000 µl, -, Transparent, Beckman®, Sherwood®, Lancer®, Bulk
779,00 Kč
1 * 1.000 KS
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • Tips in racks come with snap-on lid
  • Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
  • Containers and tips are autoclavable to +121 °C for 20 minutes
  • Balené:
    Bulk
  • Sterilní:
    -
  • Objem:
    250 - 1000 µl
  • Sekvence:
    42
  • Druh špičky:
    Standard
  • Barva špičky:
    Transparent
  • Pro použití s:
    Beckman®, Sherwood®, Lancer®