Orders LinkContactUsLinkComponent
Racks, Plexiglas for tubes
Racks, Plexiglas for tubes
12 (2×6), 28 mm, 255×90×80 mm
Catalog # 212-6350
Supplier:   KARTELL
Racks, Plexiglas for tubes
Catalog # 212-6350
Supplier:  KARTELL
Racks, Plexiglas for tubes
Racks, Plexiglas for tubes
12 (2×6), 28 mm, 255×90×80 mm
Price
1 300,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Plexiglas (PMMA)
  • Two tiers
  • Smaller holes in the lower tier to hold test tubes securely
Specifications
  • Pro zkumavku Ø:
    28 mm
  • Poč. otvorů:
    12 (2×6)
  • Š×H×V:
    255×90×80 mm