Orders LinkContactUsLinkComponent
Filter discs
Filter discs
03, 70 mm
Catalog # 519-5003
Supplier:   KARTELL
Filter discs
Catalog # 519-5003
Supplier:  KARTELL
Filter discs
Filter discs
03, 70 mm
Price
275,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
HDPE filter discs for use between filter paper and perforated Büchner plate to prevent clogging of filter holes.
Specifications
  • Ø:
    70 mm
  • Sekvence:
    03