Büchner funnels
40 ml, 42,5 mm
Katalogové číslo 519-2361
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
PP, white.
- Robust and lightweight
- Top and bottom can be separated for easy cleaning
For filter paper or membrane filtration discs.
Specifikace
- Kapacita:40 ml
- Pro filtr Ø:42,5 mm