Orders LinkContactUsLinkComponent
Drip closures
Drip closures
02, Screw cap with 40 mm nozzle, GL 18, Long spout dropping bottles
Katalogové číslo 217-0491
Dodavatel:   KARTELL
Drip closures
Katalogové číslo 217-0491
Dodavatel:  KARTELL
Drip closures
Drip closures
02, Screw cap with 40 mm nozzle, GL 18, Long spout dropping bottles
Přejít na:
Price
20,80 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
HDPE, natural colour.
  • Good chemical resistance, good temperature resistance
  • Available in two lengths
  • Attached cap
For narrow-neck bottles with round thread in accordance with DIN 168.
Specifikace
  • Určeno pro:
    Long spout dropping bottles
  • Závit:
    GL 18
  • Popis:
    Screw cap with 40 mm nozzle
  • Sekvence:
    02