Orders LinkContactUsLinkComponent
Drip closures
Drip closures
05, Screw cap with 19 mm nozzle, GL 32, Short spout dropping bottles
Katalogové číslo 217-0490
Dodavatel:   KARTELL
Drip closures
Katalogové číslo 217-0490
Dodavatel:  KARTELL
Drip closures
Drip closures
05, Screw cap with 19 mm nozzle, GL 32, Short spout dropping bottles
Přejít na:
Price
27,80 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
HDPE, natural colour.
  • Good chemical resistance, good temperature resistance
  • Available in two lengths
  • Attached cap
For narrow-neck bottles with round thread in accordance with DIN 168.
Specifikace
  • Určeno pro:
    Short spout dropping bottles
  • Závit:
    GL 32
  • Popis:
    Screw cap with 19 mm nozzle
  • Sekvence:
    05