Volumetric flasks, class B
1000 ml, 19/26, 0,80 ± ml
Katalogové číslo 612-3010
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
PMP (TPX®), transparent, with stopper.
- Autoclavable
- For working temperatures up to +170 °C
- With NS stoppers
Specifikace
- NZ:19/26
- Kapacita:1000 ml
- Tolerance:0,80 ± ml