Orders LinkContactUsLinkComponent
Erlenmeyer flasks, narrow neck, with standard socket
125 ml, 25 ml, 14/23 NS
  214-0133
 :   KARTELL
Erlenmeyer flasks, narrow neck, with standard socket
  214-0133
 :  KARTELL
Erlenmeyer flasks, narrow neck, with standard socket
Erlenmeyer flasks, narrow neck, with standard socket
125 ml, 25 ml, 14/23 NS
112,00 Kč
1 * 1 KS
PP, transparent.
  • Very good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
With moulded graduations. Autoclavable. Supplied without stoppers.
  • Interval stupnice:
    25 ml
  • Velikost objímky:
    14/23 NS
  • Kapacita:
    125 ml