Orders LinkContactUsLinkComponent
Forceps, blunt
Forceps, blunt
Stainless steel forceps, straigt, blunt, 250 mm
Catalog # BOCH1006
Supplier:   BOCHEM
Forceps, blunt
Catalog # BOCH1006
Supplier:  BOCHEM
Forceps, blunt
Forceps, blunt
Stainless steel forceps, straigt, blunt, 250 mm
Price
232,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel ¹⁸/₁₀.
Specifications
  • Popis:
    Stainless steel forceps, straigt, blunt
  • Délka:
    250 mm