Orders LinkContactUsLinkComponent
Immersion baskets, round
02, Hostalen basket with longer straps, S 40, S 40/H, 200×95×57 mm
Catalog # 142-2147
Supplier:   ELMA SCHMIDBAUER
Immersion baskets, round
Catalog # 142-2147
Supplier:  ELMA SCHMIDBAUER
Immersion baskets, round
Immersion baskets, round
02, Hostalen basket with longer straps, S 40, S 40/H, 200×95×57 mm
Price
715,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Určeno pro:
    S 40, S 40/H
  • Popis:
    Hostalen basket with longer straps
  • Sekvence:
    02
  • Š×H×V:
    200×95×57 mm