ODOUR REMOVAL ELEMENT EMF20 PK5 1 * 5 KS
Katalogové číslo EDWAA22304077
Dodavatel: EDWARDS HIGH VACUUM METTLER
ODOUR REMOVAL ELEMENT EMF20 PK5 1 * 5 KS
Katalogové číslo EDWAA22304077
Dodavatel: EDWARDS HIGH VACUUM METTLER
Frequently Bought Together
[EN]MAINBOARD 1 * 1 KS
Dodavatel: MAPADA
Accessories for 5702 / 5702 R / 5702 RH
Dodavatel: EPPENDORF
[EN]CABLE SET 230V 1 * 1 KS
Dodavatel: MEMMERT
Míchačka magnetická, atexMIXdrive
Dodavatel: 2MAG
[EN]FILTER MILLEX FG50 W 1/4INCH HOSE-CL 1 * 1 KS
Dodavatel: MEMMERT
[EN]MAINS FILTER MAX.1X250V FN2010-3-06 1 * 1 KS
Dodavatel: NABERTHERM
[DE]VERVANGPATROON OLIEMISTILF.EMF20 1 * 1 KS
Dodavatel: EDWARDS HIGH VACUUM METTLER
[EN]CONSTANT HEAD AND DRAIN, LARGE 1 * 1 KS
Dodavatel: SHERWOOD SCIENTIFIC INSTRUMENT
[EN]HEATING ELEMENT 1 * 1 KS
Dodavatel: VWR Collection
[EN]ADAPTER FOR STACKING JARS EASYFIT 12 1 * 1 KS
Dodavatel: Retsch GmbH
[EN]GAS BALLAST TUBE 56,4 MM LONG 1 * 1 KS
Dodavatel: VACUUBRAND
[EN]WASHER F/TRANSF.TOR 1-25µL 1 * 3 KS
Dodavatel: Brand
[EN]HOUSING I-TRIANGLE MB2 VDL SEALED 1 * 1 KS
Dodavatel: Binder
[EN]MOTOR FAN 35 WATT 230/50 1 * 1 KS
Dodavatel: LABCONCO
[EN]BATTERIE CR 1220 3V / 35MAH 1 * 1 KS
Dodavatel: Binder