Orders LinkContactUsLinkComponent
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
with polypropylene tip, 2 mm
Katalogové číslo 661-1181
Dodavatel:   COWIE
Stopcocks, straight through, with tip
Katalogové číslo 661-1181
Dodavatel:  COWIE
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
with polypropylene tip, 2 mm
Přejít na:
Price
615,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Made of PTFE.
  • Available with PP or glass tip
  • Very good chemical resistance
  • Very good temperature resistance
Specifikace
  • Ø otvoru:
    2 mm
  • Popis:
    with polypropylene tip