Horizontal developing chambers
Chamber for 200×100 mm plates
552-0048
: CAMAG
Horizontal developing chambers allows the number of samples to be doubled when compared to a vertical tank. Plates are developed from opposite sides simultaneously towards the middle.
- Choice of two configurations: sandwich or tank
- Suitable for all types of solvents
- Available in two plates sizes
- Popis:Chamber for 200×100 mm plates