Orders LinkContactUsLinkComponent
Flat bottom chambers
06, Chamber for 200×200 mm plates, without lid
  552-0022
 :   CAMAG
Flat bottom chambers
  552-0022
 :  CAMAG
Flat bottom chambers
Flat bottom chambers
06, Chamber for 200×200 mm plates, without lid
8 900,00 Kč
1 * 1 KS
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
  • Popis:
    Chamber for 200×200 mm plates, without lid
  • Sekvence:
    06
Accessories
  •  
    Sekvence
    Popis
  • 552-0079
    12
    Saturation pads
    1 * 100 KS - 6 630,00 Kč