Orders LinkContactUsLinkComponent
Flat bottom chambers
Flat bottom chambers
05, Chamber for 200×200 mm plates, with glass lid
Katalogové číslo 552-0015
Dodavatel:   CAMAG
Flat bottom chambers
Katalogové číslo 552-0015
Dodavatel:  CAMAG
Flat bottom chambers
Flat bottom chambers
05, Chamber for 200×200 mm plates, with glass lid
Price
10 160,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
Specifikace
  • Popis:
    Chamber for 200×200 mm plates, with glass lid
  • Sekvence:
    05
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Saturation pads
    Saturation pads
    552-0079
    12
    Přihlásit se pro dostupnost
    1 * 100 KS - 6 630,00 Kč