Flat bottom chambers
03, Chamber for 100×100 mm plates, without lid
Katalogové číslo 552-0010
Dodavatel: CAMAG
Přejít na:
Product Details & Documents
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
Specifikace
- Popis:Chamber for 100×100 mm plates, without lid
- Sekvence:03
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Saturation pads