Orders LinkContactUsLinkComponent
Flat bottom chambers
04, Chamber for 100×100 mm plates, with stainless steel lid
Katalogové číslo 552-0009
Dodavatel:   CAMAG
Flat bottom chambers
Katalogové číslo 552-0009
Dodavatel:  CAMAG
Flat bottom chambers
Flat bottom chambers
04, Chamber for 100×100 mm plates, with stainless steel lid
Price
5 950,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
Specifikace
  • Popis:
    Chamber for 100×100 mm plates, with stainless steel lid
  • Sekvence:
    04
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Saturation pads
    Saturation pads
    552-0079
    12
    Přihlásit se pro dostupnost
    1 * 100 KS - 6 630,00 Kč