Dichlormethan 99+% stabilised, extra pure
5096, Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Dichlormethan 99+% stabilised, extra pure
Catalog # ACRO124050025
Supplier: Thermo Fisher Scientific
CAS Number: 75-09-2
Jump to:
Product Details & Documents
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93
- Merck Index:12,06140
- Boiling Pt:39,8 °C (1013 hPa)
- Melting Pt:–97 °C
- Density:1,325 g/cm³
- Storage Temperature:Ambient
- EINECS:200-838-9
- UN:1593
- ADR:6.1,III
- :Stabilised with ethanol
- :Density: 1,322 g/cm³ (20 °C)
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Stabilizer:Approx. 0.2 % Ethanol
- Residue after evaporation:≤0.05 %
Specifications
- Typ balení:Glass bottle
- Bal.:2,5 L
- Sekvence:5096



