Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
 

Dichlormethan 99+% stabilised, extra pure

5037, Glass bottle, 1 L
  ACRO124050010
 
 
Dichlormethan 99+% stabilised, extra pure
  ACRO124050010
 :  75-09-2
Dichlormethan 99+% stabilised, extra pure
Dichlormethan 99+% stabilised, extra pure

5037, Glass bottle, 1 L

 :  75-09-2
 
1 540,00 Kč
   
1 * 1 L
 
 :  Methylene chloride, DCM
  •  :75-09-2
  •  :CH₂Cl₂
  •  :MFCD00000881
  •  :84.93
  •  :12,06140
  •  :39,8 °C (1013 hPa)
  •  :–97 °C
  •  :1,325 g/cm³
  •  :Ambient
  •  :200-838-9
  •  :1593
  •  :6.1,III
  •  :Stabilised with ethanol
  •  :Density: 1,322 g/cm³ (20 °C)

 

 

 

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥99.0 %
  • Stabilizer:
    Approx. 0.2 % Ethanol
  • Residue after evaporation:
    ≤0.05 %

 

  • Typ balení:
    Glass bottle
  • Bal.:
    1 L
  • Sekvence:
    5037