Accessories
Conductivity flow cell, titanium/EPDM/fluoro plastics, 14 μl volume, with M6 connection
Katalogové číslo 18-1114-43
Dodavatel: Cytiva
Přejít na:
Product Details & Documents
Documents
Specifikace
- Popis:Conductivity flow cell, titanium/EPDM/fluoro plastics, 14 μl volume, with M6 connection