Orders LinkContactUsLinkComponent
Baumé hydrometers
Baumé hydrometers
23, 50 - 60, 0,1, 300 mm
Catalog # ALLA0100FG060/15QP
Supplier:   Alla France
Baumé hydrometers
Catalog # ALLA0100FG060/15QP
Supplier:  Alla France
Baumé hydrometers
Baumé hydrometers
23, 50 - 60, 0,1, 300 mm
Price
1 260,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Baumé hydrometers calibrated at 15 °C.
Specifications
  • Délka:
    300 mm
  • Sekvence:
    23
  • Dělení:
    0,1
  • Rozsah:
    50 - 60