Orders LinkContactUsLinkComponent
Bottle Carriers
Bottle Carriers
Bottle carrier, HDPE, Šedá, Up to six bottles, 95 mm
Katalogové číslo BURK0361-0006
Dodavatel:   BURKLE
Bottle Carriers
Katalogové číslo BURK0361-0006
Dodavatel:  BURKLE
Bottle Carriers
Bottle Carriers
Bottle carrier, HDPE, Šedá, Up to six bottles, 95 mm
Přejít na:
Price
468,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Made of PP or HDPE.
  • Bottle transport container (PP) for up to 6 bottles, bottle transport box for up to 8 bottles
  • Bottle transport box (HDPE): suitable for food; With impermeable base suitable as a collecting basin
Documents
Specifikace
  • Určeno pro:
    Up to six bottles
  • Barva:
    Šedá
  • Popis:
    Bottle carrier, HDPE
  • Max. Ø nádoby:
    95 mm