Orders LinkContactUsLinkComponent
Hose clips, worm-threaded
Hose clips, worm-threaded
06, 25 - 40 mm
Catalog # BURK8677-2540
Supplier:   BURKLE
Hose clips, worm-threaded
Catalog # BURK8677-2540
Supplier:  BURKLE
Hose clips, worm-threaded
Hose clips, worm-threaded
06, 25 - 40 mm
Price
528,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Specifications
  • Upínací šířka:
    25 - 40 mm
  • Sekvence:
    06