Orders LinkContactUsLinkComponent
Spider evaporators
Spider evaporators
Spider with evaporating flasks, SJ29/32, glass, 50 ml, For simultaneous distillation from 5×50 ml evaporating flasks with STJ 24/29
Katalogové číslo 531-1319
Dodavatel:   BUCHI
Spider evaporators
Katalogové číslo 531-1319
Dodavatel:  BUCHI
Spider evaporators
Spider evaporators
Spider with evaporating flasks, SJ29/32, glass, 50 ml, For simultaneous distillation from 5×50 ml evaporating flasks with STJ 24/29
Přejít na:
Price
43 530,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifikace
  • Popis:
    Spider with evaporating flasks, SJ29/32, glass, 50 ml
  • Typ:
    For simultaneous distillation from 5×50 ml evaporating flasks with STJ 24/29