Orders LinkContactUsLinkComponent
Rotary evaporating flasks
Rotary evaporating flasks
52, 250 ml, 29/32 NS, non coated
Katalogové číslo 531-1813
Dodavatel:   BUCHI
Rotary evaporating flasks
Katalogové číslo 531-1813
Dodavatel:  BUCHI
Rotary evaporating flasks
Rotary evaporating flasks
52, 250 ml, 29/32 NS, non coated
Přejít na:
Price
2 580,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® R-210/215 rotary evaporators. Pear-shaped with NS 29.2/32 ground joint socket.
Specifikace
  • Velikost objímky:
    29/32 NS
  • Type:
    non coated
  • Kapacita:
    250 ml
  • Sekvence:
    52