Receiving flasks
2000 ml, uncoated, KS 35/20
Katalogové číslo 531-1836
Dodavatel: BUCHI
Přejít na:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Specifikace
- Zásuvka:KS 35/20
- Type:uncoated
- Kapacita:2000 ml