Stirring rods
Stirring rod, round ended, with steel core, PTFE, 6 mm, 300 mm
Catalog # BRND136325
Supplier: Brand
Jump to:
Product Details & Documents
Stirring rods.
Specifications
- Ø:6 mm
- Popis:Stirring rod, round ended, with steel core
- Délka:300 mm
- Materiál:PTFE