Orders LinkContactUsLinkComponent
Stoppers, PTFE
Stoppers, PTFE
Bílá, 12/21
Catalog # BOHLH936-03
Supplier:   Bohlender
Stoppers, PTFE
Catalog # BOHLH936-03
Supplier:  Bohlender
Stoppers, PTFE
Stoppers, PTFE
Bílá, 12/21
Price
262,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are easily removable and can be used without grease.
  • PTFE
  • Temperature resistance from −200 to + 250 °C
  • Chemical resistance
  • FDA conform
The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.
Specifications
  • NZ:
    12/21
  • Barva:
    Bílá