Powder funnels
135 mm, 140 mm, 20 mm
Catalog # 221-1617
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10.
- Smooth internal walls
- Robust and strong
- Good chemical resistance, extremely high thermal resistance
With short, wide stem.
Specifications
- Vněj. Ø nálevky:135 mm
- Délka:140 mm
- Vnější Ø stonku:20 mm