Clamps
80 mm
Katalogové číslo 241-0400
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
Stainless steel.
- Jaws are covered with a layer of cork to protect clamped items
- Without bosshead for fixing to stand
- Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894
Specifikace
- Upínací šířka:80 mm