Orders LinkContactUsLinkComponent
Tripods
With welded feet, stainless steel, 140×210 mm
Catalog # BOCH5202
Supplier:   BOCHEM
Tripods
Catalog # BOCH5202
Supplier:  BOCHEM
Tripods
Tripods
With welded feet, stainless steel, 140×210 mm
Price
1 200,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Tripods made from stainless steel or tempered cast iron (zincked).
  • With welded or screwed feet
Specifications
  • Vnitř. Ø×V:
    140×210 mm
  • Popis:
    With welded feet, stainless steel