Orders LinkContactUsLinkComponent
Tripods
Tripods
With welded feet, stainless steel, 100×180 mm
Katalogové číslo BOCH5200
Dodavatel:   BOCHEM
Tripods
Katalogové číslo BOCH5200
Dodavatel:  BOCHEM
Tripods
Tripods
With welded feet, stainless steel, 100×180 mm
Přejít na:
Price
1 030,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Tripods made from stainless steel or tempered cast iron (zincked).
  • With welded or screwed feet
Specifikace
  • Vnitř. Ø×V:
    100×180 mm
  • Popis:
    With welded feet, stainless steel