Double spatulas, micro
24, 210 mm, 55 mm, 5 mm
231-1027
: BOCHEM
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- With round handle, one flat and one rounded end
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Délka:210 mm
- Sekvence:24
- Šířka čepele:5 mm
- Délka čepele:55 mm