Double spatulas, micro
09, 130 mm, 40 mm, 4 mm
Catalog # 231-0033
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- With round handle, one flat and one rounded end
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
Specifications
- Délka:130 mm
- Sekvence:09
- Šířka čepele:4 mm
- Délka čepele:40 mm