Forceps
Blunt, 115 mm
Katalogové číslo 232-0013
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
Stainless steel, PTFE coating.
- Anti-magnetic
- Very high chemical and thermal resistance
- Smooth surface prevents any contamination and makes cleaning easier
Specifikace
- Type:Blunt
- Délka:115 mm