Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Curved, extra sharp, 120 mm
Katalogové číslo 232-0010
Dodavatel:   BOCHEM
Forceps, precision
Katalogové číslo 232-0010
Dodavatel:  BOCHEM
Forceps, precision
Forceps, precision
Curved, extra sharp, 120 mm
Přejít na:
Price
252,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production etc.
  • Extremely sharp tips without grooves
  • Reverse action; closed normally, squeeze to open
Specifikace
  • Type:
    Curved, extra sharp
  • Délka:
    120 mm